1-Butyl isothiocyanate ≥98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Butyl isothiocyanate
A10935.06 A10935.14 A10935.22
A10935.06EA 945 SEK
A10935.06 A10935.14 A10935.22
1-Butyl isothiocyanate ≥98%
1-Butyl isothiocyanate
Purity: 98+% CAS No.: 592-82-5 Molecular Formula: C5H9NS Formula Weight: 115.20 Flash Point: 66°(150°F) Refractive Index: 1.5000 Odour: Pungent Boiling Point: 63-65°/11mm Beilstein Reference: 906839 UN No.: 2922 MDL No.: MFCD00004824
Formel: C₅H₉NS
MW: 115.2 g/mol
Kokpunkt: 63…65 °C (11 mmHg)
Densitet: 0.954
Flampunkt: 66 °C (150 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004824
CAS nummer: 592-82-5
EINECS: 209-770-4
UN: 2922
ADR: 8 III