BIS-TRIS ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS), 2-(Bis(2-hydroxyethyl)amino)-2-(hydroxymethyl)propane-1,3-diol, 2-[Bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
B22515.14 B22515.22 B22515.36
B22515.14EA 715 SEK
B22515.14 B22515.22 B22515.36
BIS-TRIS ≥98%
BIS-TRIS
Purity: 98+% CAS No.: 6976-37-0 Molecular Formula: C8H19NO5 Formula Weight: 209.24 Melting Point: 102-105° Beilstein Reference: 2205275
Formel: C₈H₁₉NO₅
MW: 209,24 g/mol
Smältpunkt: 102…105 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002853
CAS nummer: 6976-37-0
EINECS: 230-237-7