BIS-TRIS 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS), 2-(Bis(2-hydroxyethyl)amino)-2-(hydroxymethyl)propane-1,3-diol, 2-[Bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
163821000 163820025 163825000
ACRO163821000EA 1150 SEK
ACRO163821000 ACRO163820025 ACRO163825000
BIS-TRIS 98%
BIS-TRIS
Formel: C₈H₁₉NO₅
MW: 209,24 g/mol
Smältpunkt: 103…104 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002853
CAS nummer: 6976-37-0
EINECS: 230-237-7
ADR: 8,III
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %