1-Dimetylaminopropan-2-ol 99%
Leverantör: Thermo Scientific
Fara
Synonyms: Dimepranol
115870010 115872500
ACRO115870010EA 2375 SEK
ACRO115870010 ACRO115872500
1-Dimetylaminopropan-2-ol 99%
1-Dimetylaminopropan-2-ol
Formel: C₅H₁₃NO
MW: 103,16 g/mol
Kokpunkt: 125…127 °C (1013 hPa)
Smältpunkt: –85 °C
Densitet: 0,85 g/cm³
Flampunkt: 26 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004532
CAS nummer: 108-16-7
EINECS: 203-556-4
UN: 2734
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid (May discolor to yellow on storage)
Infrared spectrum Conforms
Refractive index 1.4183 to 1.4203 (20°C, 589 nm)
Water ≤0.5 %
GC ≥98.5 %