Tripropylamine 98%

Leverantör: Thermo Scientific
Fara

A10285.K2 A10285.AK
A10285.K2EA 2100 SEK
A10285.K2 A10285.AK
Tripropylamine 98%
Tripropylamine

Purity: 98% CAS No.: 102-69-2 Molecular Formula: C9H21N Formula Weight: 143.27 Flash Point: 34°(93°F) Refractive Index: 1.4160 Odour: Amine-like Melting Point: -93° Boiling Point: 156° UN No.: 2260 MDL No.: MFCD00009363

Formel: C₉H₂₁N
MW: 143,27 g/mol
Kokpunkt: 156 °C
Smältpunkt: –93 °C
Densitet: 0,755
Flampunkt: 34 °C (93 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009363
CAS nummer: 102-69-2
EINECS: 203-047-7
UN: 2260
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR