N,N-Dimethylformamide di-tert-butyl acetal tech. 90%, Technical Grade
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1,1-Bis(1,1-dimethylethoxy)-N,N,N-trimethylamine, 1,1-Di-tert-butoxy-N,N-dimethylmethanamine, 1,1-Di-tert-butoxytrimethylamine
L00431.06 L00431.14
L00431.06EA 2450 SEK
L00431.06 L00431.14
N,N-Dimethylformamide di-tert-butyl acetal tech. 90%, Technical Grade
N,N-Dimethylformamide di-tert-butyl acetal
Purity: 90% CAS No.: 36805-97-7 Molecular Formula: C11H25NO2 Formula Weight: 203.33 Flash Point: 33°(91°F) Refractive Index: 1.4120 Boiling Point: 75-77°/12mm Beilstein Reference: 969629 UN No.: 1993 MDL No.: MFCD00015002
Formel: C₁₁H₂₅NO₂
MW: 203.32 g/mol
Kokpunkt: 75…77 °C (12 mmHg)
Densitet: 0.850
Flampunkt: 33 °C (91 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00015002
CAS nummer: 36805-97-7
EINECS: 253-222-7
UN: 1993
ADR: 3 III