Trifluoromethanesulphonic acid 99%, extra pure
Leverantör: Thermo Scientific
Fara
Synonyms: Trifluoromethanesulphonic acid
169890010 169890100 169890500 169892500 169890011
ACRO169890010EA 860 SEK
ACRO169890010 ACRO169890100 ACRO169890500 ACRO169892500 ACRO169890011
Trifluoromethanesulphonic acid 99%, extra pure
Trifluoromethanesulphonic acid
Formel: CHF₃O₃S
MW: 150,08 g/mol
Kokpunkt: 162 °C (760 mmHg)
Smältpunkt: –40 °C
Densitet: 1,69 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00007514
CAS nummer: 1493-13-6
EINECS: 216-087-5
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.3260 to 1.3300 (20°C, 589 nm)
Titration with NaOH ≥98.5 %