Trimethylsilyl trifluoromethanesulphonate ≥99%
Leverantör: Thermo Scientific
Fara
Synonyms: Trimethylsilyl trifluoromethanesulphonate
A12535.30 A12535.09 A12535.18
A12535.30EA 14550 SEK
A12535.30 A12535.09 A12535.18
Trimethylsilyl trifluoromethanesulphonate ≥99%
Trimethylsilyl trifluoromethanesulphonate
Purity: 99% CAS No.: 27607-77-8 Molecular Formula: (CH3)3SiSO3CF3 Formula Weight: 222.26 Flash Point: 25°(77°F) Refractive Index: 1.3600 Boiling Point: 39-40°/12mm Beilstein Reference: 1868911 Merck Reference: 14,9719 UN No.: 2924 MDL No.: MFCD00000406
Formel: C₄H₉F₃O₃SSi
MW: 222,26 g/mol
Kokpunkt: 39…40 °C (12 mmHg)
Densitet: 1,225
Flampunkt: 25 °C (77 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00000406
CAS nummer: 27607-77-8
EINECS: 248-565-4
ADR: 3,III
Merck Index: 14,9719