3,3,3-Trifluoro-1-propanol ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 3,3,3-Trifluoropropan-1-ol
L16879.03 L16879.06
L16879.03EA 2225 SEK
L16879.03 L16879.06
3,3,3-Trifluoro-1-propanol ≥97%
3,3,3-Trifluoro-1-propanol
Purity: 97% CAS No.: 2240-88-2 Molecular Formula: CF3CH2CH2OH Formula Weight: 114.07 Flash Point: 37°(98°F) Refractive Index: 1.3200 Boiling Point: 100-101° Beilstein Reference: 1734777 UN No.: 1987 MDL No.: MFCD00190649
Formel: C₃H₅F₃O
MW: 114,07 g/mol
Kokpunkt: 100…101 °C
Densitet: 1,294
Flampunkt: 37 °C (98 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00190649
CAS nummer: 2240-88-2
UN: 1987
ADR: 3,III