2,2,3,3,4,4,4-Heptafluoro-1-butanol ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 1H,1H-Perfluoro-1-butanol, 2,2,3,3,4,4,4-Heptafluorobutan-1-ol, Heptafluoro-1-butanol
A10414.09 A10414.14 A10414.18
A10414.09EA 1600 SEK
A10414.09 A10414.14 A10414.18
2,2,3,3,4,4,4-Heptafluoro-1-butanol ≥98%
2,2,3,3,4,4,4-Heptafluoro-1-butanol
Purity: 98% CAS No.: 375-01-9 Molecular Formula: C4H3F7O Formula Weight: 200.06 Flash Point: 25°(77°F) Refractive Index: 1.300 Boiling Point: 95-97° Beilstein Reference: 1761907 UN No.: 1987 MDL No.: MFCD00004674
Formel: C₄H₃F₇O
MW: 200,06 g/mol
Kokpunkt: 95…97 °C
Densitet: 1,596
Flampunkt: 25 °C (77 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004674
CAS nummer: 375-01-9
EINECS: 206-782-1
UN: 1987
ADR: 3,III