Pentafluoropropionic anhydride ≥99% for GC derivatisation, Supelco®
Leverantör: Merck
Fara
Synonyms: Perfluoropropanoic anhydride, Pentafluoropropanoic anhydride, PFPA
M-Clarity™ quality level = MQ100
394904-5ML 394904-100ML 394904-10X1ML 394904-25ML
394904-5MLEA 1160 SEK
394904-5ML 394904-100ML 394904-10X1ML 394904-25ML
Pentafluoropropionic anhydride ≥99% for GC derivatisation, Supelco®
Pentafluoropropionic anhydride
Formel: C₆F₁₀O₃
MW: 310,05 g/mol
Kokpunkt: 69 °C (1013 hPa)
Densitet: 1,571 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000429
CAS nummer: 356-42-3
EINECS: 206-604-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c11de0b34bbfdca3716184820c4d52be97cf4d7&vid=5c11de0b34bbfdca3716184820c4d52be97cf4d7&prid=web&clid=SAPDAM