Pentafluoropropionic anhydride 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Perfluoropropanoic anhydride, Pentafluoropropanoic anhydride, PFPA
416930250 416930050 416931000
ACRO416930250EA 4250 SEK
ACRO416930250 ACRO416930050 ACRO416931000
Pentafluoropropionic anhydride 98%
Pentafluoropropionic anhydride
Formel: C₆F₁₀O₃
MW: 310.05 g/mol
Kokpunkt: 69 °C
Densitet: 1.57 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00000429
CAS nummer: 356-42-3
EINECS: 206-604-2
UN: 3265
ADR: 8 III
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %