1-Amino-2,2,2-trifluoroethane 99.5%
Leverantör: Thermo Fisher Scientific
Fara
303500010 303500100 303500500
ACRO303500010EA 625 SEK
ACRO303500010 ACRO303500100 ACRO303500500
1-Amino-2,2,2-trifluoroethane 99.5%
1-Amino-2,2,2-trifluoroethane
Formel: C₂H₄F₃N
Kokpunkt: 37…38 °C (760 mmHg)
Densitet: 1,24 g/cm³
Flampunkt: –16 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00008132
CAS nummer: 753-90-2
EINECS: 212-041-3
UN: 3286
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to almost colorless Liquid
Infrared spectrum Conforms
GC ≥99.4 %