Hexafluoroglutaric acid ≥98%

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: Hexafluoro-1,5-pentanedioic acid

A17848.06 A17848.14
A17848.06EA 1250 SEK
A17848.06 A17848.14
Hexafluoroglutaric acid ≥98%
Hexafluoroglutaric acid

Purity: 98% CAS No.: 376-73-8 Molecular Formula: C5H2F6O4 Formula Weight: 240.06 Melting Point: 92-97° Boiling Point: 134-138°/3mm UN No.: 3261 MDL No.: MFCD00004172

Formel: C₅H₂F₆O₄
Kokpunkt: 134…138 °C (3 mmHg)
Smältpunkt: 92…97 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004172
CAS nummer: 376-73-8
EINECS: 206-813-9
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR