2,2-Difluorosuccinic acid ≥94%
Leverantör: Thermo Scientific
Fara
Synonyms: α,α-Difluorosuccinic acid, 2,2-Difluoro-1,4-butanedioic acid
L00773.03 L00773.06
L00773.03EA 3200 SEK
L00773.03 L00773.06
2,2-Difluorosuccinic acid ≥94%
2,2-Difluorosuccinic acid
Purity: 94% CAS No.: 665-31-6 Molecular Formula: C4H4F2O4 Formula Weight: 154.07 Flash Point: 110°(230°F) Melting Point: 139-144° Beilstein Reference: 1773355 UN No.: 3261 MDL No.: MFCD00040970
Formel: C₄H₄F₂O₄
MW: 154,07 g/mol
Kokpunkt: 268 °C (1013 hPa)
Smältpunkt: 139…144 °C
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00040970
CAS nummer: 665-31-6
EINECS: 211-558-1
UN: 3261
ADR: 8,III