Pentafluoropropionic acid 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Perfluoropropionic acid, Pentafluoropropanoic acid, Perfluoropropanoic acid, 2,2,3,3,3-Pentafluoropropionic acid
416920100 416920500 416922500
ACRO416920100EA 985 SEK
ACRO416920100 ACRO416920500 ACRO416922500
Pentafluoropropionic acid 97%
Pentafluoropropionic acid
Formel: C₃HF₅O₂
MW: 164.03 g/mol
Kokpunkt: 96 °C (760 mmHg)
Densitet: 1.576 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00004170
CAS nummer: 422-64-0
EINECS: 207-021-6
UN: 3265
ADR: 8 III
Order Now
Specifikation testresultat
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %
GC ≥97.5 %