Methyl trifluoroacetate 99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Trifluoroacetic methyl ester
243410250 243411000
ACRO243410250EA 1450 SEK
ACRO243410250 ACRO243411000
Methyl trifluoroacetate 99%
Methyl trifluoroacetate
Formel: C₃H₃F₃O₂
Kokpunkt: 43…43,5 °C (760 mmHg)
Smältpunkt: –78 °C
Densitet: 1,273 g/cm³
Flampunkt: –7 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000417
CAS nummer: 431-47-0
EINECS: 207-074-5
UN: 2924
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥98.5 %