Ethyl trifluoroacetate 99%
Leverantör: Thermo Scientific
Fara
Synonyms: Trifluoroacetic ethyl ester
119020250 119021000 119025000 119020025
ACRO119020250EA 730 SEK
ACRO119020250 ACRO119021000 ACRO119025000 ACRO119020025
Ethyl trifluoroacetate 99%
Ethyl trifluoroacetate
Formel: C₄H₅F₃O₂
MW: 142,08 g/mol
Kokpunkt: 60…62 °C (760 mmHg)
Smältpunkt: –78 °C
Densitet: 1,194 g/cm³
Flampunkt: –1 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000419
CAS nummer: 383-63-1
EINECS: 206-851-6
UN: 3272
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥98.5 %
HPLC ≥94 %