Trifluoroacetamide ≥97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2,2,2-Trifluoroacetamide
A14451.14 A14451.22 A14451.36
A14451.14EA 915 SEK
A14451.14 A14451.22 A14451.36
Trifluoroacetamide ≥97%
Trifluoroacetamide
Purity: 97% CAS No.: 354-38-1 Molecular Formula: CF3CONH2 Formula Weight: 113.04 Melting Point: 70-75° Boiling Point: 162-164° MDL No.: MFCD00008008
Formel: C₂H₂F₃NO
MW: 113.04 g/mol
Kokpunkt: 162…164 °C
Smältpunkt: 70…75 °C
Densitet: 1 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008008
CAS nummer: 354-38-1
EINECS: 206-559-9