EDTA (Etylendiamintetraättiksyra) ≥99.4% ACS
Leverantör: Thermo Fisher Scientific
Varning
11931.22 11931.36 11931.A4
11931.22EA 835 SEK
11931.22 11931.36 11931.A4
EDTA (Etylendiamintetraättiksyra) ≥99.4% ACS
EDTA (Etylendiamintetraättiksyra)
Purity: 99.4+% CAS No.: 60-00-4 Molecular Formula: C10H16N2O8 Formula Weight: 292.24 Physical Form: Powder Appearance: White Melting Point: ca 245° dec. Beilstein Reference: 1716295 Merck Reference: 14,3517 MDL No.: MFCD00003541
Chelating agent
Formel: C₁₀H₁₆N₂O₈
Kokpunkt: 614 °C (1013 hPa)
Smältpunkt: ∼ 245 °C (dec.)
Densitet: 0,86 g/cm³ (20 °C)
Flampunkt: >100 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003541
CAS nummer: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 14,3517
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM