Oxalsyra, anhydrous 98
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Ethanedioic acid
44410.18 44410.30
44410.18EA 995 SEK
44410.18 44410.30
Oxalsyra, anhydrous 98
Oxalsyra
Purity: 98% CAS No.: 144-62-7 Molecular Formula: C2H2O4 Formula Weight: 90.04 Physical Form: Crystalline Density: 1.9 Appearance: White pH: 1.3 Melting Point: ca 185° dec. Beilstein Reference: 385686 Merck Reference: 14,6911 UN No.: 3261 MDL No.: MFCD00002573
Formel: C₂H₂O₄
MW: 90.04 g/mol
Kokpunkt: 365
Smältpunkt: ∼ 185 °C (dec.)
Densitet: 1.9
Storage Temperature: Omgivande
MDL Number: MFCD00002573
CAS nummer: 144-62-7
EINECS: 205-634-3
UN: 3261
ADR: 8 III
Merck Index: 14,6911