Azelaic acid 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Azelaic acid,, 1,7-Heptanedicarboxylic acid, 1,9-Nonanedioic acid
401520250 401521000 401525000
ACRO401520250EA 545 SEK
ACRO401520250 ACRO401521000 ACRO401525000
Azelaic acid 98%
Azelaic acid
Formel: C₉H₁₆O₄
MW: 188,22 g/mol
Kokpunkt: 237 °C (20hPa)
Smältpunkt: 98…103 °C
Densitet: 1,0287 g/cm³ (20 °C)
Flampunkt: 215 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004432
CAS nummer: 123-99-9
EINECS: 204-669-1
Merck Index: 13,00908
Order Now
Specifikation testresultat
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
GC ≥97.5 %