Suberic acid zS, Sigma-Aldrich®
Leverantör: Merck
Varning
Synonyms: 1,8-Octanedioic acid, Octanedioic acid, 1,6-Hexanedicarboxylic acid
MQ200
8.18800.0100 8.18800.0500
8.18800.0100EA 659 SEK
8.18800.0100 8.18800.0500
Suberic acid zS, Sigma-Aldrich®
Suberic acid
Formel: C₈H₁₄O₄
MW: 174.2 g/mol
Kokpunkt: 361
Smältpunkt: 139…142 °C
Densitet: 1.354 g/cm³ (21 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004428
CAS nummer: 505-48-6
EINECS: 208-010-9
Merck Index: 12,09031
Order Now
Specifikation testresultat
Solubility 1.6 g/l (20 °C)
Melting Point 139 - 142 °C
Molar Mass 174.2 g/mol
Bulk Density 700 kg/m3
Boiling Point 229 - 231 °C (20 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 210 °C
pH 3.2 (1.6 g/l, H2O, 20 °C)
Ignition Point 430 °C
Synonyms Suberic acid
Hill Formula C8H14O4
Chemical Formula HOOC(CH2)6COOH