Glutarsyra ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1,5-Pentanedioic acid
A14595.14 A14595.22 A14595.36
A14595.14EA 580 SEK
A14595.14 A14595.22 A14595.36
Glutarsyra ≥99%
Glutarsyra
Purity: 99% CAS No.: 110-94-1 Molecular Formula: C5H8O4 Formula Weight: 132.12 Melting Point: 95-99° Boiling Point: 200°/20mm Merck Reference: 14,4473 MDL No.: MFCD00004410
Formel: C₅H₈O₄
MW: 132.12 g/mol
Kokpunkt: 200 °C (20 mmHg)
Smältpunkt: 95…99 °C
Densitet: 1.429
Storage Temperature: Omgivande
MDL Number: MFCD00004410
CAS nummer: 110-94-1
EINECS: 203-817-2
Merck Index: 13,04486