Bärnstenssyra ≥99%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 1,4-Butanedioic acid
A12084.0C A12084.0E A12084.36
A12084.0CEA 10125 SEK
A12084.0C A12084.0E A12084.36
Bärnstenssyra ≥99%
Bärnstenssyra
Purity: 99+% CAS No.: 110-15-6 Molecular Formula: HO2CCH2CH2CO2H Formula Weight: 118.09 Flash Point: 206°(402°F) Melting Point: 186-188° Boiling Point: 235° Beilstein Reference: 1754069 Merck Reference: 14,8869 MDL No.: MFCD00002789
Formel: HOOCCH₂CH₂COOH
MW: 118.09 g/mol
Kokpunkt: 235 °C
Smältpunkt: 186…188 °C
Densitet: 1.56
Flampunkt: 206 °C (402 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00002789
CAS nummer: 110-15-6
EINECS: 203-740-4
UN: 3261
ADR: 8 III
Merck Index: 13,08953
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd381e6e0e4cbde676a92c9e8d6aacfac635ade&vid=0b14e1b7f82b03468fc82d224a0ee894952eb200&prid=web&clid=SAPDAM