1,18-Octadecanedioic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Octadecanedioic acid, 1,16-Hexadecanedicarboxylic acid, EMEROX 118
O0222-1G
O0222-25G
O0222-5G
TCIAO0222-1GEA
615
SEK
TCIAO0222-1G
TCIAO0222-25G
TCIAO0222-5G
1,18-Octadecanedioic acid ≥98.0% (by GC, titration analysis)
1,18-Octadecanedioic acid
Formel:
C₁₈H₃₄O₄ MW: 314,47 g/mol |
MDL Number:
MFCD00142369 CAS nummer: 871-70-5 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 124.0 to 127.0 deg-C |
Solubility in hot Methanol | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...