4-Aminobutyric acid 97%
Leverantör: Thermo Scientific
Synonyms: GABA, γ-Aminobutyric acid
A11016.0E A11016.22 A11016.36
A11016.0EEA 14250 SEK
A11016.0E A11016.22 A11016.36
4-Aminobutyric acid 97%
4-Aminobutyric acid
Purity: 97% CAS No.: 56-12-2 Molecular Formula: C4H9NO2 Formula Weight: 103.12 Odour: Odourless Melting Point: ca 195° dec. Beilstein Reference: 906818 Merck Reference: 14,430 MDL No.: MFCD00008226
Formel: C₄H₉NO₂
MW: 103,12 g/mol
Kokpunkt: 248 °C (1013 hPa)
Smältpunkt: ∼ 195 °C (dec.)
Densitet: 1,21 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008226
CAS nummer: 56-12-2
EINECS: 200-258-6
Merck Index: 14,430