Cobalt(II) acetate Co ≥ 32%, vattenfri
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Cobaltous acetate, Cobalt acetate, Acetic acid cobalt (II) salt, Cobalt diacetate
23138.14 23138.22
23138.14EA 3300 SEK
23138.14 23138.22
Cobalt(II) acetate Co ≥ 32%, vattenfri
Cobalt(II) acetate
CAS No.: 71-48-7 Molecular Formula: C4H6CoO4 Formula Weight: 177.02 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Pink Odour: Odourless Melting Point: 298° dec. Solubility: Soluble in water Merck Reference: 14,2433 UN No.: 3077 MDL No.: MFCD00008689
Formel: C₄H₆CoO₄
MW: 177.02 g/mol
Smältpunkt: 298 °C (dec.)
Densitet: 1.7755 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008689
CAS nummer: 71-48-7
EINECS: 200-755-8
UN: 3077
ADR: 9 III
Merck Index: 14,2433