Koppar (II) acetat, vattenfri 99%, ren
Leverantör: Thermo Scientific
Fara
Synonyms: Acetic acid copper (II) salt, Cupric acetate, Copper diacetate
405370500 405370010 405372500
ACRO405370500EA 2950 SEK
ACRO405370500 ACRO405370010 ACRO405372500
Koppar (II) acetat, vattenfri 99%, ren
Copper(II) acetate
Formel: CuCH₃CO₂
MW: 181,64 g/mol
Kokpunkt: 240 °C (1013 hPa)
Smältpunkt: 115 °C
Densitet: 1,9 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008690
CAS nummer: 142-71-2
EINECS: 205-553-3
UN: 3260
ADR: 8,II
Merck Index: 12,02690
Order Now
Specifikation testresultat
Appearance Green to blue Crystalline powder
Infrared spectrum Conforms
Titration with Na2S2O3 ≥98.5 %