Tallium(I)acetat 99%
Leverantör: Thermo Scientific
Fara
Synonyms: Acetic acid thallium (I) salt, Thallous acetate
420770250 420771000 420775000
ACRO420770250EA 2650 SEK
ACRO420770250 ACRO420771000 ACRO420775000
Tallium(I)acetat 99%
Tallium(I)acetat
Formel: C₂H₃O₂Tl
MW: 263,43 g/mol
Kokpunkt: 1457 °C (1013 hPa)
Smältpunkt: 128…130 °C
Densitet: 3,68 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00013045
CAS nummer: 563-68-8
EINECS: 209-257-5
UN: 1707
ADR: 6.1,II
Merck Index: 12,09392
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
Titration with NaOH ≥96.0 %