Natriumoktanoat, EMPROVE® EXPERT Ph. Eur., NF suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Varning
Synonyms: Octanoic acid sodium acid, Sodium caprylate, Caprylic acid sodium acid, Sodium n-Octanoate
8.17081.9025 8.17081.1000 8.17081.9010 8.17081.9011
8.17081.9025EA 0 SEK
8.17081.9025 8.17081.1000 8.17081.9010 8.17081.9011
Natriumoktanoat, EMPROVE® EXPERT Ph. Eur., NF suitable for use as excipient, SAFC®
Natriumoktanoat
Formel: C₈H₁₅NaO₂
Smältpunkt: 225…355 °C
Densitet: >0,9949 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00058511
CAS nummer: 1984-06-1
EINECS: 217-850-5