Lithium stearate 95%
Leverantör: Thermo Fisher Scientific
Synonyms: Stearic acid lithium salt, Lithium Stearate
413340250 413341000
ACRO413340250EA 450 SEK
ACRO413340250 ACRO413341000
Lithium stearate 95%
Lithium stearate
Formel: C₁₈H₃₅LiO₂
MW: 290.42 g/mol
Smältpunkt: 220 °C
Storage Temperature: Omgivande
MDL Number: MFCD00042032
CAS nummer: 4485-12-5
EINECS: 224-772-5
Order Now
Specifikation testresultat
Appearance White Powder
Infrared spectrum Conforms
Titration with HClO4 ≥94 %
GC ≥97.5 %