Natriumformiat ≥98%
Leverantör: Thermo Scientific
Synonyms: Formic acid sodium salt
A17813.0B A17813.0I A17813.30
A17813.0BEA 800 SEK
A17813.0B A17813.0I A17813.30
Natriumformiat ≥98%
Natriumformiat
Purity: 98% CAS No.: 141-53-7 Molecular Formula: HCO2Na Formula Weight: 68.01 Melting Point: 259-262° Beilstein Reference: 3595134 Merck Reference: 14,8621 MDL No.: MFCD00013101
Formel: HCO₂Na
MW: 68,01 g/mol
Smältpunkt: 259…262 °C
Densitet: 1,92
Storage Temperature: Omgivande
MDL Number: MFCD00013101
CAS nummer: 141-53-7
EINECS: 205-488-0
REACH: 01-2119486468-21
Merck Index: 14,8621