Natriumformiat 99%
Leverantör: Thermo Fisher Scientific
148230010 148230051
ACRO148230010EA 860 SEK
ACRO148230010 ACRO148230051
Natriumformiat 99%
Natriumformiat
Formel: HCO₂Na
Kokpunkt: 360 °C (760 mmHg)
Smältpunkt: 258…261 °C
Densitet: 1,919 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00013101
CAS nummer: 141-53-7
EINECS: 205-488-0
REACH: 01-2119486468-21
Merck Index: 13,08694
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 253 to 261 °C
Water ≤0.5 %
Titration with HClO4 ≥98.5 %