Natriumacetat, vattenfri ≥99.997%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Acetic acid sodium salt
11005.06 11005.14 11005.22
11005.06EA 1050 SEK
11005.06 11005.14 11005.22
Natriumacetat, vattenfri ≥99.997%
Natriumacetat
Purity: 99.997% CAS No.: 127-09-3 Molecular Formula: NaOOCCH3 Formula Weight: 82.03 Physical Form: Powder Density: 1.528 pH: 7,5-9 Melting Point: 324° Beilstein Reference: 3595639 Merck Reference: 14,8571 MDL No.: MFCD00012459
Formel: H₃CCOONa
MW: 82.03 g/mol
Kokpunkt: 881.4
Smältpunkt: 324
Densitet: 1.528
Flampunkt: >250 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00012459
CAS nummer: 127-09-3
EINECS: 204-823-8
REACH: 01-2119485123-42
Merck Index: 14,8571