Sodium butyrate 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Butyric acid sodium salt, Sodium Butyrate
263190050 263190250 263191000 263195000
ACRO263190050EA 485 SEK
ACRO263190050 ACRO263190250 ACRO263191000 ACRO263195000
Sodium butyrate 98%
Sodium butyrate
Formel: C₄H₇NaO₂
Smältpunkt: 250…253 °C
Densitet: 1,324 g/cm³ (30 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002816
CAS nummer: 156-54-7
EINECS: 205-857-6
Order Now
Specifikation testresultat
Appearance White Powder
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %