Triethyl citrate ≥99%
Leverantör: Thermo Fisher Scientific
L12639.30 L12639.14 L12639.0B
L12639.30EA 770 SEK
L12639.30 L12639.14 L12639.0B
Triethyl citrate ≥99%
Triethyl citrate
Purity: 99% CAS No.: 77-93-0 Molecular Formula: C12H20O7 Formula Weight: 276.29 Flash Point: 176°(348°F) Refractive Index: 1.4420 Melting Point: -55° Boiling Point: 126-127°/1mm Beilstein Reference: 1801199 Merck Reference: 14,2326 MDL No.: MFCD00009201
Formel: C₁₂H₂₀O₇
Kokpunkt: 126…127 °C (1 mmHg)
Smältpunkt: –55 °C
Densitet: 1,137
Flampunkt: 176 °C (348 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009201
CAS nummer: 77-93-0
EINECS: 201-070-7
Merck Index: 14,2326