Dietyladipat 99%
Leverantör: Thermo Scientific
Synonyms: NSC 19160, Adipic acid diethyl ester, 1,6-Hexanedioic acid diethyl ester, NSC 3363
407171000 407175000
ACRO407171000EA 500 SEK
ACRO407171000 ACRO407175000
Dietyladipat 99%
Dietyladipat
Formel: C₁₀H₁₈O₄
MW: 202,25 g/mol
Kokpunkt: 132 °C (15 mmHg)
Smältpunkt: –18 °C
Densitet: 1 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009215
CAS nummer: 141-28-6
EINECS: 205-477-0
Merck Index: 13,00163
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4280 (20°C, 589 nm)
GC ≥98.5 %