Dimethyl glutarate 98%
Leverantör: Thermo Fisher Scientific
Synonyms: 1,5-Pentanedioic acid dimethyl ester, Glutaric acid dimethyl ester
116231000 116235000
ACRO116231000EA 680 SEK
ACRO116231000 ACRO116235000
Dimethyl glutarate 98%
Dimethyl glutarate
Formel: C₇H₁₂O₄
MW: 160.17 g/mol
Kokpunkt: 211
Smältpunkt: –37 °C
Densitet: 1.087 g/cm³
Flampunkt: 103 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008468
CAS nummer: 1119-40-0
EINECS: 214-277-2
Merck Index: 13,04486
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4236 to 1.4256 (20°C, 589 nm)
GC ≥97.5 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %