Isopropyl myristate ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Tetradecanoic acid isopropyl ester, Myristic acid isopropyl ester, Isopropyl Myristate, Isopropyl tetradecanoate, Tetradecylic acid isopropyl ester
A18111.AP A18111.AE A18111.0F
A18111.APEA 2100 SEK
A18111.AP A18111.AE A18111.0F
Isopropyl myristate ≥98%
Isopropyl myristate
Purity: 98% CAS No.: 110-27-0 Molecular Formula: CH3(CH2)12CO2CH(CH3)2 Formula Weight: 270.46 Flash Point: 152°(306°F) Physical Form: Liquid Density: 0.85 Appearance: Colourless Boiling Point: 191-194°/20mm MDL No.: MFCD00008982
Formel: C₁₇H₃₄O₂
Kokpunkt: 191…194 °C (20 mmHg)
Smältpunkt: 0…1 °C
Densitet: 0,85
Flampunkt: 152 °C (306 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008982
CAS nummer: 110-27-0
EINECS: 203-751-4
Merck Index: 13,05235