Methyl stearate ≥99%
Leverantör: Thermo Fisher Scientific
Synonyms: Stearic acid methyl ester, Methyl octadecanoate
A13265.06 A13265.14
A13265.06EA 1275 SEK
A13265.06 A13265.14
Methyl stearate ≥99%
Methyl stearate
Purity: 99% CAS No.: 112-61-8 Molecular Formula: CH3(CH2)16CO2CH3 Formula Weight: 298.51 Flash Point: 110°(230°F) Physical Form: Cryst powder Melting Point: 38-42° Boiling Point: 180-182/4mm Beilstein Reference: 1786213 Merck Reference: 14,8804 MDL No.: MFCD00009005
Formel: CH₃(CH₂)₁₆CO₂CH₃
Kokpunkt: 180…182 °C (4 mmHg)
Smältpunkt: 38…42 °C
Densitet: 1,007 g/cm³ (20 °C)
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009005
CAS nummer: 112-61-8
EINECS: 203-990-4
Merck Index: 14,8804