Metylformiat ≥97% may cont. up to ca 3% methanol
Leverantör: Thermo Fisher Scientific
|
Fara
|
Purity: 97%
CAS No.: 107-31-3
Molecular Formula: C2H4O2
Formula Weight: 60.05
Storage Temperature: 2°C to 8°C
Flash Point: -26°(-14°F)
Physical Form: Clear liquid
Density: 0.972
Refractive Index: 1.3440
Appearance: Colourless
Melting Point: -100°
Boiling Point: 32-33°
Beilstein Reference: 1734623
Merck Reference: 14,6077
UN No.: 1243
MDL No.: MFCD00003291
Formel:
C₂H₄O₂ Kokpunkt: 32…33 °C Smältpunkt: –100 °C Densitet: 0,972 Flampunkt: –26 °C (–14 °F) Storage Temperature: Kylskåp |
MDL Number:
MFCD00003291 CAS nummer: 107-31-3 EINECS: 203-481-7 UN: 1243 ADR: 3,I Merck Index: 14,6077 |