Methyl hexanoate ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester
A10757.0B A10757.18 A10757.30
A10757.0BEA 4400 SEK
A10757.0B A10757.18 A10757.30
Methyl hexanoate ≥99%
Methyl hexanoate
Purity: 99% CAS No.: 106-70-7 Molecular Formula: CH3(CH2)4CO2CH3 Formula Weight: 130.19 Flash Point: 45°(113°F) Refractive Index: 1.4050 Melting Point: -71° Boiling Point: 150-151° Beilstein Reference: 1744683 UN No.: 3272 MDL No.: MFCD00009510
Formel: C₇H₁₄O₂
MW: 130,19 g/mol
Kokpunkt: 150…151 °C
Smältpunkt: –71 °C
Densitet: 0,885
Flampunkt: 45 °C (113 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009510
CAS nummer: 106-70-7
EINECS: 203-425-1
UN: 3272
ADR: 3,III