Ethyl laurate
Leverantör: Cayman Chemical
Synonyms: Dodecanoic acid ethyl ester, Ethyl dodecanoate, Ethyl Laurate, Dodecylic acid ethyl ester, Lauric acid ethyl ester
10008203-100 10008203-500 10008203-1
CAYM10008203-100EA 435 SEK
CAYM10008203-100 CAYM10008203-500 CAYM10008203-1
Ethyl laurate
Ethyl laurate
Formel: C₁₄H₂₈O₂
MW: 228,38 g/mol
Kokpunkt: 269 °C (1013 hPa)
Smältpunkt: –2 °C
Densitet: 0,86 g/cm³ (20 °C)
Flampunkt: 138 °C
Storage Temperature: Omgivande
MDL Number: MFCD00015065
CAS nummer: 106-33-2
EINECS: 203-386-0
Merck Index: 13,03851