Formanilide ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: N-Phenyl-formamide
A12472.22 A12472.14 A12472.36
A12472.22EA 1650 SEK
A12472.22 A12472.14 A12472.36
Formanilide ≥98%
Formanilide
Purity: 98% CAS No.: 103-70-8 Molecular Formula: C7H7NO Formula Weight: 121.14 Flash Point: 110°(230°F) Melting Point: 45-49° Boiling Point: 271° Beilstein Reference: 906934 Merck Reference: 14,4238 MDL No.: MFCD00003276
Formel: C₇H₇NO
MW: 121,14 g/mol
Kokpunkt: 271 °C
Smältpunkt: 45…49 °C
Densitet: 1,140
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00003276
CAS nummer: 103-70-8
EINECS: 203-136-0
Merck Index: 14,4238