Formanilide 99+%
Leverantör: Thermo Scientific
Synonyms: N-Phenyl-formamide
158420050 158421000 158425000
ACRO158420050EA 400 SEK
ACRO158420050 ACRO158421000 ACRO158425000
Formanilide 99+%
Formanilide
Formel: C₇H₇NO
MW: 121,14 g/mol
Kokpunkt: 271 °C (1013 hPa)
Smältpunkt: 47…49 °C
Densitet: 1,14 g/cm³
Flampunkt: >112 °C
MDL Number: MFCD00003276
CAS nummer: 103-70-8
EINECS: 203-136-0
Merck Index: 13,04262
Order Now
Specifikation testresultat
Appearance White to light grey-beige Crystals
Infrared spectrum Conforms
Melting point 46 to 49 °C
GC ≥99.0 %
UV at 280 nm A: ≤0.05
pH 4.0 to 5.0 (10 % in water)
Titration with NaOH ≥98.5 %