2'-Methylacetanilide ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: o-Acetotoluidide, o-Acetotoluidid, N-(o-Tolyl)acetamide, N-(2-Methylphenyl)acetamide
A13235.14 A13235.22
A13235.14EA 1525 SEK
A13235.14 A13235.22
2'-Methylacetanilide ≥98%
2'-Methylacetanilide
Purity: 98+% CAS No.: 120-66-1 Molecular Formula: C9H11NO Formula Weight: 149.19 Melting Point: 109-112° Boiling Point: 296° Merck Reference: 14,74 MDL No.: MFCD00014961
Formel: C₉H₁₁NO
MW: 149,19 g/mol
Kokpunkt: 296 °C
Smältpunkt: 109…112 °C
Densitet: 1,160
Storage Temperature: Omgivande
MDL Number: MFCD00014961
CAS nummer: 120-66-1
EINECS: 204-414-4
Merck Index: 14,74