Acetohydrazide 95%
Leverantör: Thermo Scientific
Varning
Synonyms: Acetic hydrazide, Acethydrazide
102350250 102351000 102355000
ACRO102350250EA 855 SEK
ACRO102350250 ACRO102351000 ACRO102355000
Acetohydrazide 95%
Acetohydrazide
Formel: C₂H₆N₂O
MW: 74,08 g/mol
Kokpunkt: 129 °C (18 mmHg)
Smältpunkt: 57…60 °C
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007610
CAS nummer: 1068-57-1
EINECS: 213-948-7
UN: 1325
ADR: 4.1,III
Order Now
Specifikation testresultat
Appearance White Moist crystals and/or chunks
Infrared spectrum Conforms
Assay ≥94 % (on dry substance)
Loss on drying (50°C, 3 h) Balance
Gel strength ≥750 g/cm² (1 % gel)
Gel point 34.5 to 37.5°C (1.5 % gel)
Electroendosmosis 0.16 to 0.19 (-Mr)
Ash ≤0.5 %
Sulfate (SO4) ≤0.25 %