Urea, EMPROVE® EXPERT BP, Ph. Eur., ACS, JP, USP BiopharmaceuticalProduction, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
1.37030.9029 1.37030.5000 1.37030.9012 1.37030.9026
1.37030.9029EA 0 SEK
1.37030.9029 1.37030.5000 1.37030.9012 1.37030.9026
Urea, EMPROVE® EXPERT BP, Ph. Eur., ACS, JP, USP BiopharmaceuticalProduction, SAFC®
Urea
Formel: NH₂CONH₂
MW: 60.06 g/mol
Kokpunkt: 197
Smältpunkt: 132.7
Densitet: 1.34 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008022
CAS nummer: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM