Formamid, EMPLURA®, Supelco®
Leverantör: Merck
|
Fara
|
Synonyms:
Carbamaldehyde
M-Clarity™ quality level = MQ200
1.04008.2500
1.04008.9025
1.04008.2500EA
1515
SEK
1.04008.2500
1.04008.9025
Formamid, EMPLURA®, Supelco®
Formamid
Formel:
HCONH₂ MW: 45,04 g/mol Kokpunkt: 210 °C (1013 hPa) Smältpunkt: 2 °C Densitet: 1,1334 g/cm³ (20 °C) Flampunkt: 79 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00007941 CAS nummer: 75-12-7 EINECS: 200-842-0 Merck Index: 13,04261 |
|
Learn more
![](/m-se.vwr.com/sv_SE/images/about_vwr_275x315.jpg)